siRNA Details
| Gene Symbol: Miip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025365.2 | NP_001020536.2 | 3 | 884 | ||
| GenBank mRNA | AK133976.1 | - | 5 | 1036 | ||
| AK170224.1 | - | 3 | 677 | |||
| BC017525.1 | - | 3 | 627 | 4910951 | ||
| BC024069.1 | - | 3 | 666 | 5097088 | ||
| FJ618908.1 | - | 3 | 884 | |||

Miip
Genomic Map