siRNA Details
| Gene Symbol: Desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134095.2 | NP_598856.1 | 6 | 1404 | ||
| GenBank mRNA | AK003172.1 | - | 1 | 124 | ||
| AK005963.1 | - | 6 | 1360 | |||
| AK016347.1 | - | 6 | 1310 | |||
| AK131703.1 | - | 6 | 1357 | |||
| AK146155.1 | - | 6 | 1428 | |||
| AK170731.1 | - | 7 | 1673 | |||
| AK216343.1 | - | 1 | 160 | |||
| AK218907.1 | - | 1 | 155 | |||
| BC022097.1 | - | 5 | 1359 | |||
| BC056972.1 | - | 6 | 1404 | 6811343 | ||
| BC071205.1 | - | 6 | 1296 | 6515068 | ||

Desi1
Genomic Map