siRNA Details
| Gene Symbol: Nelfe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045863.1 | NP_001039328.1 | 6, 7 | 618 | ||
| NM_001045864.1 | NP_001039329.1 | 6, 7 | 704 | |||
| NM_138580.2 | NP_613046.2 | 6, 7 | 492 | |||
| GenBank mRNA | AK011663.1 | - | 6, 7 | 476 | ||
| AK012389.1 | - | 6, 7 | 509 | |||
| AK131947.1 | - | 6, 7 | 476 | |||
| AK147011.1 | - | 6, 7 | 600 | |||
| BC034867.1 | - | 6, 7 | 444 | 1495547 | ||
| CN525174.1 | - | 6, 7 | 447 | |||
| DV662511.1 | - | 3, 4 | 237 | |||

Nelfe
Genomic Map