siRNA Details
| Gene Symbol: 1700088E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138581.2 | NP_613047.1 | 7 | 1042 | ||
| GenBank mRNA | AK006539.1 | - | 7 | 1023 | ||
| AK030004.1 | - | 1378 | ||||
| BC068203.1 | - | 5 | 904 | |||
| BC096672.1 | - | 5 | 957 | 5117646 | ||

1700088E04Rik
Genomic Map