siRNA Details
| Gene Symbol: Spg21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138584.2 | NP_613050.1 | 9 | 1618 | ||
| GenBank mRNA | AK011985.1 | - | 9 | 1585 | ||
| AK149930.1 | - | 9 | 1592 | |||
| AK150215.1 | - | 9 | 1619 | |||
| AK153284.1 | - | 9 | 1580 | |||
| BC099434.1 | - | 8 | 1383 | 30714184 | ||
| BC132377.1 | - | 9 | 1595 | 40130654 | ||

Spg21
Genomic Map