siRNA Details
| Gene Symbol: Zfp330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145600.1 | NP_663575.1 | 9, 10 | 762 | ||
| GenBank mRNA | AK029489.1 | - | 8, 9 | 587 | ||
| AK037748.1 | - | 3, 4 | 2693 | |||
| AK050534.1 | - | 9, 10 | 772 | |||
| BC008086.1 | - | 9, 10 | 762 | 3586161 | ||
| BC030675.1 | - | 9, 10 | 760 | 1068780 | ||
| BC037025.1 | - | 5, 6 | 2119 | |||
| BC083081.1 | - | 9, 10 | 796 | 6510396 | ||

Zfp330
Genomic Map