siRNA Details
| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145705.3 | NP_663751.2 | 9 | 1443 | ||
| GenBank mRNA | AF214013.1 | - | 9 | 1431 | ||
| AF518764.1 | - | 9 | 1408 | |||
| AK011154.1 | - | 9 | 1411 | |||
| AK016456.1 | - | 9 | 1377 | |||
| AK028952.1 | - | 8 | 1568 | |||
| AK090324.1 | - | 9 | 1410 | |||
| AK169480.1 | - | 3 | 1561 | |||
| AK205383.1 | - | 2 | 107 | |||
| AK205420.1 | - | 2 | 104 | |||
| AK214714.1 | - | 3 | 150 | |||
| BC017142.1 | - | 4 | 1859 | |||
| BC030347.1 | - | 9 | 1369 | 5003804 | ||

Tinf2
Genomic Map