siRNA Details
| Gene Symbol: Wipi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145940.2 | NP_666052.1 | 5 | 538 | ||
| GenBank mRNA | AK031672.1 | - | 5 | 547 | ||
| AK034907.1 | - | 5 | 582 | |||
| AK045131.1 | - | 5 | 547 | |||
| AK050495.1 | - | 5 | 546 | |||
| AK159608.1 | - | 5 | 567 | |||
| AK170563.1 | - | 5 | 582 | |||
| AK186984.1 | - | 3 | 210 | |||
| AK203625.1 | - | 3 | 210 | |||
| BC024811.1 | - | 5 | 506 | |||
| BC024883.1 | - | 5 | 502 | |||
| BC025560.1 | - | 5 | 538 | 5322999 | ||

Wipi1
Genomic Map