siRNA Details
| Gene Symbol: Amot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290274.1 | NP_001277203.1 | 6 | 1733 | ||
| NM_153319.3 | NP_695231.3 | 6 | 1733 | |||
| GenBank mRNA | AF461135.1 | - | 6 | 1137 | ||
| BC057638.1 | - | 6 | 1188 | 2631191 | ||
| CK392840.1 | - | 3 | 179 | |||

Amot
Genomic Map