siRNA Details
| Gene Symbol: Tubgcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290824.1 | NP_001277753.1 | 5 | 657 | ||
| NM_153387.3 | NP_700436.1 | 5 | 657 | |||
| GenBank mRNA | AK016563.1 | - | 5 | 700 | ||
| AK038921.1 | - | 5 | 616 | |||
| AK045305.1 | - | 5 | 550 | |||
| AK046132.1 | - | 5 | 587 | |||
| AK050250.1 | - | 5 | 658 | |||
| AK051517.1 | - | 5 | 558 | |||
| AK152984.1 | - | 5 | 616 | |||
| AK153171.1 | - | 5 | 616 | |||
| AK190869.1 | - | 2 | 88 | |||
| BC029106.1 | - | 5 | 621 | 3711922 | ||

Tubgcp4
Genomic Map