siRNA Details
| Gene Symbol: Stbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175096.3 | NP_780305.1 | 2 | 311 | ||
| GenBank mRNA | AK050416.1 | - | 2 | 312 | ||
| BC052666.1 | - | 2 | 272 | |||
| BC118615.1 | - | 3 | 380 | 40107322 | ||
| BC118661.1 | - | 2 | 294 | 40107321 | ||

Stbd1
Genomic Map