siRNA Details
| Gene Symbol: Ccdc69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177471.4 | NP_803422.2 | 6 | 565 | ||
| GenBank mRNA | AK008760.1 | - | 6 | 536 | ||
| AK079810.1 | - | 6 | 566 | |||
| AK135942.1 | - | 6 | 542 | |||
| AK170686.1 | - | 6 | 515 | |||
| BC096763.1 | - | 6 | 523 | 30053993 | ||

Ccdc69
Genomic Map