siRNA Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170855.1 | NP_001164326.1 | 10 | 3084 | ||
| NM_178872.4 | NP_849203.2 | 10 | 3085 | |||
| GenBank mRNA | AK033754.1 | - | 1 | 888 | ||
| BC058106.1 | - | 10 | 3004 | 6836197 | ||

Trim36
Genomic Map