siRNA Details
| Gene Symbol: Gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181649.6 | NP_857632.4 | 5 | 615 | ||
| GenBank mRNA | AK009084.1 | - | 5 | 502 | ||
| AK148331.1 | - | 5 | 617 | |||
| BC047998.1 | - | 5 | 529 | 6511097 | ||
| BC092522.1 | - | 5 | 542 | 30648550 | ||
| BC138310.1 | - | 5 | 510 | 8861330 | ||
| BY741452.1 | - | 5 | 616 | |||

Gpatch11
Genomic Map