siRNA Details
| Gene Symbol: Ceacam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007675.4 | NP_031701.3 | 4 | 1027 | ||
| GenBank mRNA | AK018529.1 | - | 4 | 1007 | ||
| BC003346.1 | - | 4 | 1048 | 3481822 | ||
| L38422.1 | - | 4 | 991 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam10| Gene Symbol: Ceacam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007675.4 | NP_031701.3 | 4 | 1027 | ||
| GenBank mRNA | AK018529.1 | - | 4 | 1007 | ||
| BC003346.1 | - | 4 | 1048 | 3481822 | ||
| L38422.1 | - | 4 | 991 | |||