siRNA Details
| Gene Symbol: Psg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007677.2 | NP_031703.1 | 2 | 563 | ||
| GenBank mRNA | BC100345.1 | - | 2 | 560 | 30715425 | |
| CN703708.1 | - | 2 | 563 | |||
| M83344.1 | - | 2 | 579 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psg17| Gene Symbol: Psg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007677.2 | NP_031703.1 | 2 | 563 | ||
| GenBank mRNA | BC100345.1 | - | 2 | 560 | 30715425 | |
| CN703708.1 | - | 2 | 563 | |||
| M83344.1 | - | 2 | 579 | |||