siRNA Details
| Gene Symbol: Map4k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252200.1 | NP_001239129.1 | 20 | 2641 | ||
| NM_001252201.1 | NP_001239130.1 | 18 | 2248 | |||
| NM_001252202.1 | NP_001239131.1 | 19 | 2410 | |||
| NM_008696.2 | NP_032722.2 | 19 | 2479 | |||
| GenBank mRNA | AK129191.1 | - | 19 | 2310 | ||
| AK139433.1 | - | 1 | 104 | |||
| BC141109.1 | - | 18 | 1982 | 9055721 | ||
| BC145390.1 | - | 20 | 2375 | 9053948 | ||
| BC145391.1 | - | 19 | 2143 | 9053949 | ||
| CX231827.1 | - | 3 | 252 | |||
| U88984.1 | - | 19 | 2159 | |||

Map4k4
Genomic Map