siRNA Details
| Gene Symbol: Tarbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253795.1 | NP_001240724.1 | 5 | 732 | ||
| NM_009319.3 | NP_033345.2 | 7 | 1005 | |||
| GenBank mRNA | AK143188.1 | - | 6 | 904 | ||
| AK144205.1 | - | 5 | 570 | |||
| BC002028.1 | - | 7 | 718 | 3489376 | ||
| BC053432.1 | - | 2 | 1505 | |||
| U79962.1 | - | 7 | 738 | |||

Tarbp2
Genomic Map