siRNA Details
| Gene Symbol: Tcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289468.1 | NP_001276397.1 | 3 | 1094 | ||
| NM_009337.3 | NP_033363.1 | 4 | 1164 | |||
| GenBank mRNA | AF031956.1 | - | 4 | 1141 | ||
| AK149361.1 | - | 3 | 1076 | |||
| AK163387.1 | - | 4 | 1148 | |||
| BC052336.1 | - | 4 | 1145 | 30053730 | ||
| DV660020.1 | - | 1 | 267 | |||
| Y15376.1 | - | 4 | 1141 | |||

Tcl1
Genomic Map