siRNA Details
| Gene Symbol: Prss40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009356.2 | NP_033382.2 | 4 | 693 | ||
| GenBank mRNA | AB008911.1 | - | 4 | 676 | ||
| AK006524.1 | - | 2 | 303 | |||
| AK132909.1 | - | 4 | 723 | |||
| BC145989.1 | - | 4 | 708 | 40131292 | ||

Prss40
Genomic Map