siRNA Details
| Gene Symbol: Tgfb1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289550.1 | NP_001276479.1 | 11 | 1377 | ||
| NM_001289551.1 | NP_001276480.1 | 11 | 1373 | |||
| NM_001289552.1 | NP_001276481.1 | 10 | 1234 | |||
| NM_001289553.1 | NP_001276482.1 | 10 | 1261 | |||
| GenBank mRNA | AK003170.1 | - | 11 | 1211 | ||
| AK041233.1 | - | 11 | 1206 | |||
| AK085042.1 | - | 10 | 1104 | |||
| AK087114.1 | - | 11 | 1208 | |||
| AK158409.1 | - | 10 | 1086 | |||
| BC002049.1 | - | 11 | 1211 | 3489459 | ||
| BC056362.1 | - | 10 | 2486 | 6816930 | ||
| DQ143891.1 | - | 10 | 1262 | |||
| DQ143892.1 | - | 9 | 1048 | |||
| DQ143893.1 | - | 9 | 1055 | |||
| DQ143894.1 | - | 9 | 1050 | |||
| DQ143895.1 | - | 9 | 1466 | |||
| DQ143896.1 | - | 9 | 1427 | |||
| DQ143897.1 | - | 9 | 1505 | |||
| DQ143898.1 | - | 9 | 1454 | |||
| DQ143899.1 | - | 8 | 1581 | |||
| DQ143900.1 | - | 10 | 1274 | |||
| L22482.1 | - | 10 | 1338 | |||

Tgfb1i1
Genomic Map