siRNA Details
| Gene Symbol: Tnfsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009403.3 | NP_033429.1 | 4 | 632 | ||
| GenBank mRNA | AK030916.1 | - | 4 | 633 | ||
| AK157270.1 | - | 4 | 385 | |||
| BC117093.1 | - | 4 | 571 | 40126412 | ||
| BC117097.1 | - | 4 | 571 | 40126416 | ||
| L09754.1 | - | 4 | 519 | |||

Tnfsf8
Genomic Map