siRNA Details
| Gene Symbol: IFRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006764.4 | NP_006755.4 | 3 | 551 | ||
| GenBank mRNA | AK074763.1 | - | 3 | 368 | ||
| BC001327.1 | - | 3 | 378 | 3456988 | ||
| BC001676.2 | - | 3 | 363 | 2963821 | ||
| BC007265.1 | - | 3 | 386 | 3138466 | ||
| BC007437.2 | - | 3 | 411 | 2961581 | ||
| BI825477.1 | - | 3 | 551 | |||
| BT019999.1 | - | 3 | 230 | |||
| U09585.1 | - | 3 | 228 | |||
| Y12395.1 | - | 6 | 728 | |||

IFRD2
Genomic Map