siRNA Details
| Gene Symbol: Tpd52l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009413.1 | NP_033439.1 | 7 | 1020 | ||
| GenBank mRNA | AF004428.1 | - | 7 | 1020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpd52l1| Gene Symbol: Tpd52l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009413.1 | NP_033439.1 | 7 | 1020 | ||
| GenBank mRNA | AF004428.1 | - | 7 | 1020 | ||