siRNA Details
Gene Symbol: Tpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009417.2 | NP_033443.1 | 6 | 631 | ||
GenBank mRNA | AK047843.1 | - | 6 | 610 | ||
X60703.1 | - | 6 | 631 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009417.2 | NP_033443.1 | 6 | 631 | ||
GenBank mRNA | AK047843.1 | - | 6 | 610 | ||
X60703.1 | - | 6 | 631 |