siRNA Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204071.1 | NP_001191000.1 | 6 | 453 | ||
| NM_009420.2 | NP_033446.1 | 6 | 471 | |||
| GenBank mRNA | AK018856.1 | - | 6 | 402 | ||
| BC049615.1 | - | 6 | 423 | 6742033 | ||
| CJ046590.1 | - | 6 | 394 | |||
| M25533.1 | - | 6 | 423 | |||

Crisp2
Genomic Map