siRNA Details
| Gene Symbol: Traf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290413.1 | NP_001277342.1 | 7 | 748 | ||
| NM_009422.3 | NP_033448.2 | 7 | 727 | |||
| GenBank mRNA | AF027570.1 | - | 6 | 639 | ||
| AK038136.1 | - | 7 | 676 | |||
| AK052934.1 | - | 7 | 676 | |||
| AK151653.1 | - | 7 | 665 | |||
| AK152174.1 | - | 7 | 695 | |||
| BC003801.1 | - | 6 | 653 | 3492839 | ||
| BC060625.1 | - | 5 | 2684 | |||
| L35303.1 | - | 7 | 664 | |||

Traf2
Genomic Map