siRNA Details
| Gene Symbol: Traf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290413.1 | NP_001277342.1 | 11 | 2903 | ||
| NM_009422.3 | NP_033448.2 | 11 | 2882 | |||
| GenBank mRNA | AK052934.1 | - | 11 | 2831 | ||
| AK159585.1 | - | 2829 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf2| Gene Symbol: Traf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290413.1 | NP_001277342.1 | 11 | 2903 | ||
| NM_009422.3 | NP_033448.2 | 11 | 2882 | |||
| GenBank mRNA | AK052934.1 | - | 11 | 2831 | ||
| AK159585.1 | - | 2829 | ||||