siRNA Details
| Gene Symbol: Traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155434.1 | - | 3 | 565 | ||
| BC060705.1 | - | 4 | 786 | 6409657 | ||
| D84655.1 | - | 4 | 639 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf6| Gene Symbol: Traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155434.1 | - | 3 | 565 | ||
| BC060705.1 | - | 4 | 786 | 6409657 | ||
| D84655.1 | - | 4 | 639 | |||