siRNA Details
| Gene Symbol: Tst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009437.4 | NP_033463.1 | 2 | 1032 | ||
| GenBank mRNA | BC005644.1 | - | 2 | 1056 | 3257034 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tst| Gene Symbol: Tst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009437.4 | NP_033463.1 | 2 | 1032 | ||
| GenBank mRNA | BC005644.1 | - | 2 | 1056 | 3257034 | |