siRNA Details
| Gene Symbol: Tgoln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009443.3 | NP_033469.1 | 4 | 2830 | ||
| GenBank mRNA | AK076586.1 | - | 4 | 2831 | ||
| AK133559.1 | - | 4 | 2577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgoln1| Gene Symbol: Tgoln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009443.3 | NP_033469.1 | 4 | 2830 | ||
| GenBank mRNA | AK076586.1 | - | 4 | 2831 | ||
| AK133559.1 | - | 4 | 2577 | |||