siRNA Details
Gene Symbol: Ube2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009456.2 | NP_033482.1 | 4 | 435 | ||
GenBank mRNA | AJ130961.1 | - | 4 | 424 | ||
AK007265.1 | - | 4 | 443 | |||
AK152522.1 | - | 4 | 436 | |||
AK159876.1 | - | 1 | 24 | |||
AK165620.1 | - | 4 | 416 | |||
AK167847.1 | - | 4 | 423 | |||
BC049075.1 | - | 4 | 422 | |||
BC093503.1 | - | 4 | 414 | 4216580 | ||
BC106149.1 | - | 4 | 413 | 4502101 | ||
X97042.1 | - | 4 | 435 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108847, XM_008768927, FQ221006, BC167087.
Panther Classification:
Gene Ontology Categories:
