siRNA Details
| Gene Symbol: Ube2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169576.1 | NP_001163047.1 | 2, 3 | 570 | ||
| NM_009459.3 | NP_033485.1 | 2, 3 | 570 | |||
| GenBank mRNA | AK131688.1 | - | 2, 3 | 548 | ||
| AK161414.1 | - | 2, 3 | 554 | |||
| BC008517.1 | - | 2, 3 | 376 | 3600233 | ||
| U19854.1 | - | 2, 3 | 299 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001172127.

Ube2h
Genomic Map