siRNA Details
| Gene Symbol: Ube2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169576.1 | NP_001163047.1 | 5 | 841 | ||
| NM_001169577.1 | NP_001163048.1 | 5 | 417 | |||
| NM_009459.3 | NP_033485.1 | 7 | 934 | |||
| GenBank mRNA | AK131688.1 | - | 7 | 912 | ||
| AK161414.1 | - | 5 | 825 | |||
| BC008517.1 | - | 7 | 740 | 3600233 | ||
| U19854.1 | - | 7 | 663 | |||

Ube2h
Genomic Map