siRNA Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009469.3 | NP_033495.2 | 25, 26 | 3170 | ||
| GenBank mRNA | AF053756.1 | - | 25, 26 | 2862 | ||
| AF072370.1 | - | 25, 26 | 2853 | |||
| AK122357.1 | - | 24, 25 | 2672 | |||
| AK148317.1 | - | 25, 26 | 3163 | |||
| AK154939.1 | - | 25, 26 | 3187 | |||
| AK156534.1 | - | 25, 26 | 3147 | |||
| BC023281.1 | - | 1, 2 | 96 | |||
| BC030850.1 | - | 4, 5 | 642 | |||
| BC053062.1 | - | 25, 26 | 2830 | |||
| BC057121.1 | - | 25, 26 | 3170 | 6834534 | ||
| BC059835.1 | - | 25, 26 | 3139 | 6406755 | ||

Ulk1
Genomic Map