siRNA Details
| Gene Symbol: Umod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278605.1 | NP_001265534.1 | 2, 3 | 236 | ||
| NM_009470.5 | NP_033496.1 | 2, 3 | 292 | |||
| GenBank mRNA | AK085460.1 | - | 2, 3 | 286 | ||
| AK144065.1 | - | 2, 3 | 208 | |||
| AK164688.1 | - | 2, 3 | 208 | |||
| AK165507.1 | - | 2, 3 | 152 | |||
| BB843740.1 | - | 2, 3 | 237 | |||
| BC012973.1 | - | 1, 2 | 130 | 4210940 | ||

Umod
Genomic Map