siRNA Details
| Gene Symbol: Uros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135314.1 | - | 8, 9 | 1315 | ||
| AK145902.1 | - | 9, 10 | 873 | |||
| AK148116.1 | - | 9, 10 | 1010 | |||
| BC068161.1 | - | 9, 10 | 816 | 30358504 | ||
| U04439.1 | - | 8, 9 | 737 | |||
| U16216.1 | - | 9, 10 | 878 | |||
| U18867.2 | - | 9, 10 | 882 | |||

Uros
Genomic Map