siRNA Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009483.1 | NP_033509.1 | 29 | 4807 | ||
| GenBank mRNA | AK162600.1 | - | 13 | 2074 | ||
| AK164248.1 | - | 10 | 1409 | |||
| BC053433.1 | - | 29 | 4807 | 6516368 | ||
| BC075703.1 | - | 20 | 3700 | |||

Kdm6a
Genomic Map