siRNA Details
| Gene Symbol: Uty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009484.2 | NP_033510.2 | 12 | 1321 | ||
| GenBank mRNA | AF057367.1 | - | 12 | 1301 | ||
| AK142344.1 | - | 14 | 1405 | |||
| AK142512.1 | - | 12 | 1237 | |||
| AK155922.1 | - | 13 | 1201 | |||
| AK162410.1 | - | 11 | 1029 | |||
| AK220463.1 | - | 13 | 1361 | |||
| BC053061.1 | - | 12 | 1304 | |||

Uty
Genomic Map