siRNA Details
| Gene Symbol: Vamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009498.4 | NP_033524.1 | 5 | 954 | ||
| GenBank mRNA | AK051272.1 | - | 5 | 955 | ||
| AK051691.1 | - | 5 | 949 | |||
| AK165947.1 | - | 5 | 903 | |||
| AK168413.1 | - | 5 | 943 | |||
| AK169056.1 | - | 5 | 952 | |||
| AK169087.1 | - | 5 | 946 | |||
| AK213255.1 | - | 1 | 35 | |||
| BC060045.1 | - | 5 | 930 | 5690213 | ||

Vamp3
Genomic Map