siRNA Details
| Gene Symbol: Wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009516.3 | NP_033542.2 | 2 | 891 | ||
| GenBank mRNA | AK011212.1 | - | 2 | 667 | ||
| AK039572.1 | - | 2 | 207 | |||
| BC006852.1 | - | 2 | 910 | 3156953 | ||
| D30743.1 | - | 2 | 892 | |||

Wee1
Genomic Map