siRNA Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271757.1 | NP_001258686.1 | 5 | 1862 | ||
| NM_001271758.1 | NP_001258687.1 | 5 | 1805 | |||
| NM_009525.3 | NP_033551.2 | 5 | 2059 | |||
| GenBank mRNA | AK013218.1 | - | 5 | 2030 | ||
| AK049558.1 | - | 5 | 1830 | |||
| BC010775.1 | - | 5 | 1805 | 4211437 | ||
| BC051406.1 | - | 5 | 1990 | 6438917 | ||

Wnt5b
Genomic Map