siRNA Details
| Gene Symbol: Xpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009531.2 | NP_033557.2 | 9 | 1769 | ||
| GenBank mRNA | AB071144.1 | - | 9 | 1706 | ||
| AK004713.1 | - | 9 | 1709 | |||
| AK028595.1 | - | 9 | 1707 | |||
| AK166981.1 | - | 9 | 1704 | |||
| AK170975.1 | - | 9 | 1701 | |||
| BC052725.1 | - | 9 | 1731 | 6812198 | ||
| U27398.1 | - | 9 | 1680 | |||
| U40005.1 | - | 8 | 1508 | |||

Xpc
Genomic Map