siRNA Details
| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009537.3 | NP_033563.2 | 1 | 542 | ||
| GenBank mRNA | BC055899.1 | - | 1 | 520 | 3156776 | |
| M73963.1 | - | 1 | 542 | |||
| M74590.1 | - | 1 | 482 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yy1| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009537.3 | NP_033563.2 | 1 | 542 | ||
| GenBank mRNA | BC055899.1 | - | 1 | 520 | 3156776 | |
| M73963.1 | - | 1 | 542 | |||
| M74590.1 | - | 1 | 482 | |||