siRNA Details
| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009537.3 | NP_033563.2 | 5 | 1338 | ||
| GenBank mRNA | AK076149.1 | - | 6 | 951 | ||
| AK166542.1 | - | 6 | 676 | |||
| BC055899.1 | - | 5 | 1316 | 3156776 | ||
| M73963.1 | - | 5 | 1338 | |||
| M74590.1 | - | 5 | 1278 | |||

Yy1
Genomic Map