siRNA Details
| Gene Symbol: Rnf103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146898.1 | - | 4 | 1817 | ||
| AK168644.1 | - | 4 | 1998 | |||
| BC137624.1 | - | 5 | 1355 | 8860643 | ||
| D76445.1 | - | 4 | 1829 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053438, XM_006236653, BC100629, AF306394.

Rnf103
Genomic Map