siRNA Details
| Gene Symbol: Zbtb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009547.2 | NP_033573.1 | 3 | 1215 | ||
| GenBank mRNA | AK037725.1 | - | 3 | 1336 | ||
| AK082051.1 | - | 3 | 1356 | |||
| AK136762.1 | - | 1311 | ||||
| AK145517.1 | - | 3 | 1011 | |||
| AK146201.1 | - | 3 | 1015 | |||
| BC052017.1 | - | 3 | 1215 | 5697838 | ||
| L15325.1 | - | 2 | 946 | |||

Zbtb14
Genomic Map