siRNA Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290350.1 | NP_001277279.1 | 4 | 2387 | ||
| NM_001290351.1 | NP_001277280.1 | 4 | 2378 | |||
| NM_001290353.1 | NP_001277282.1 | 4 | 2285 | |||
| NM_009554.4 | NP_033580.3 | 4 | 2375 | |||
| GenBank mRNA | AK032469.1 | - | 4 | 2257 | ||
| BC024863.1 | - | 2 | 1998 | |||

Zfp37
Genomic Map