siRNA Details
| Gene Symbol: Zfp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009555.2 | NP_033581.2 | 6 | 2807 | ||
| GenBank mRNA | AK049894.1 | - | 6 | 2814 | ||
| BC065097.1 | - | 6 | 2800 | |||
| D10632.1 | - | 2 | 2489 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp40| Gene Symbol: Zfp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009555.2 | NP_033581.2 | 6 | 2807 | ||
| GenBank mRNA | AK049894.1 | - | 6 | 2814 | ||
| BC065097.1 | - | 6 | 2800 | |||
| D10632.1 | - | 2 | 2489 | |||