siRNA Details
| Gene Symbol: Zfp92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009566.4 | NP_033592.2 | 6 | 2603 | ||
| GenBank mRNA | AK141522.1 | - | 6 | 2604 | ||
| U47104.1 | - | 6 | 2623 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp92| Gene Symbol: Zfp92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009566.4 | NP_033592.2 | 6 | 2603 | ||
| GenBank mRNA | AK141522.1 | - | 6 | 2604 | ||
| U47104.1 | - | 6 | 2623 | |||